Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256252.1 | NP_001243181.1 | 20 | 2733 | ||
NM_001256253.1 | NP_001243182.1 | 20 | 2595 | |||
NM_001256254.1 | NP_001243183.1 | 18 | 2367 | |||
NM_001256255.1 | NP_001243184.1 | 18 | 2259 | |||
NM_001974.4 | NP_001965.3 | 21 | 2790 | |||
XM_011527794.1 | XP_011526096.1 | 23 | 3066 | |||
GenBank mRNA | AK290401.1 | - | 21 | 2769 | ||
AK291518.1 | - | 20 | 2565 | |||
AK297003.1 | - | 18 | 2345 | |||
AK297011.1 | - | 18 | 2232 | |||
BC059395.1 | - | 20 | 2578 | 30349201 | ||
X81479.1 | - | 21 | 2767 |
Set Membership: |
G-protein coupled receptor |