Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003384.2 | NP_003375.1 | 3 | 313 | ||
XM_006720247.3 | XP_006720310.1 | 3 | 351 | |||
XM_011537132.1 | XP_011535434.1 | 2 | 212 | |||
XM_017021622.1 | XP_016877111.1 | 2 | 212 | |||
XM_017021624.1 | XP_016877113.1 | 3 | 351 | |||
XM_017021625.1 | XP_016877114.1 | 3 | 392 | |||
XM_017021626.1 | XP_016877115.1 | 3 | 351 | |||
GenBank mRNA | AB000449.1 | - | 3 | 282 | ||
AK290110.1 | - | 3 | 301 | |||
BC103761.1 | - | 3 | 291 | 5773877 | ||
BC112075.1 | - | 3 | 281 | 8327543 | ||
BC113510.1 | - | 3 | 281 | 8322562 | ||
BU535261.1 | - | 3 | 333 |