Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005290.3 | NP_005281.1 | 1 | 975 | ||
GenBank mRNA | AK313375.1 | - | 1 | 975 | ||
BC069437.1 | - | 1 | 840 | 7262234 | ||
BC101779.1 | - | 1 | 866 | 8069285 | ||
BC101781.1 | - | 1 | 863 | 8069287 |
Set Membership: |
G-protein coupled receptor |