Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014365.2 | NP_055180.1 | 3 | 975 | ||
GenBank mRNA | AF133207.1 | - | 3 | 812 | ||
AF191017.1 | - | 3 | 975 | |||
AF217987.1 | - | 3 | 820 | |||
AF250138.1 | - | 3 | 460 | |||
AK312501.1 | - | 3 | 828 | |||
AL136936.1 | - | 3 | 805 | |||
BC002673.2 | - | 3 | 943 | 3607117 | ||
BT006876.1 | - | 3 | 452 | |||
CR533453.1 | - | 3 | 452 |