Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000396.3 | NP_000387.1 | 8 | 1533 | ||
GenBank mRNA | AK130134.1 | - | 8 | 1350 | ||
BC016058.1 | - | 8 | 1415 | 4860926 | ||
S79895.1 | - | 8 | 1445 | |||
U13665.1 | - | 8 | 1409 | |||
U20280.1 | - | 8 | 1322 |