Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006223.3 | NP_006214.2 | 4 | 1017 | ||
NR_033187.1 | 3 | 943 | ||||
GenBank mRNA | AB009690.1 | - | 4 | 938 | ||
AF143096.1 | - | 4 | 931 | |||
BC005234.1 | - | 4 | 936 | |||
BC070288.1 | - | 4 | 936 | |||
BC111394.1 | - | 4 | 937 |