Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282425.1 | NP_001269354.1 | 4 | 798 | ||
NM_006572.5 | NP_006563.2 | 4 | 1108 | |||
GenBank mRNA | AF493902.1 | - | 4 | 899 | ||
AK301699.1 | - | 5 | 1033 | |||
AK302400.1 | - | 4 | 798 | |||
AK313672.1 | - | 4 | 902 | |||
BC036756.1 | - | 4 | 953 | 5548632 | ||
L22075.1 | - | 4 | 940 |