Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP2A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005173.3 | NP_005164.2 | 21 | 4688 | ||
NM_174953.2 | NP_777613.1 | 23 | 4847 | |||
NM_174954.2 | NP_777614.1 | 23 | 4834 | |||
NM_174955.2 | NP_777615.1 | 22 | 4776 | |||
NM_174956.2 | NP_777616.1 | 22 | 4789 | |||
NM_174957.2 | NP_777617.1 | 21 | 4685 | |||
NM_174958.2 | NP_777618.1 | 23 | 4195 | |||
XM_011523885.1 | XP_011522187.1 | 22 | 4773 | |||
XM_011523888.2 | XP_011522190.1 | 22 | 4786 | |||
GenBank mRNA | AF068221.1 | - | 11 | 2423 | ||
Z69881.1 | - | 21 | 4521 |