Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005805.5 | NP_005796.1 | 4 | 557 | ||
GenBank mRNA | AK055128.1 | - | 4 | 794 | ||
BC066336.1 | - | 4 | 403 | 5296432 | ||
U86782.1 | - | 4 | 289 |