Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001827.2 | NP_001818.1 | 1 | 118 | ||
GenBank mRNA | BC006458.1 | - | 1 | 84 | 3538689 | |
BM838320.1 | - | 1 | 118 | |||
BM841681.1 | - | 1 | 121 | |||
BT006630.1 | - | 1 | 20 | |||
CR407630.1 | - | 1 | 20 | |||
CR542118.1 | - | 1 | 20 | |||
X54942.1 | - | 1 | 115 |