Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002591.3 | NP_002582.3 | 3 | 528 | ||
GenBank mRNA | AK290802.1 | - | 3 | 506 | ||
BC023978.1 | - | 3 | 503 | 4042259 | ||
BM768055.1 | - | 3 | 531 | |||
BX648510.1 | - | 1 | 1103 | |||
DB187814.1 | - | 3 | 499 | |||
L05144.1 | - | 3 | 486 |