Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004252.4 | NP_004243.1 | 6 | 1680 | ||
GenBank mRNA | AF015926.1 | - | 6 | 1657 | ||
AF036241.1 | - | 6 | 1615 | |||
AK026581.1 | - | 6 | 1663 | |||
AK094467.1 | - | 5 | 1326 | |||
AK128474.1 | - | 7 | 1629 | |||
BC001443.2 | - | 6 | 1633 | 3138881 | ||
BC003361.1 | - | 6 | 1650 | 2900516 | ||
BC011777.1 | - | 6 | 1649 | 3640162 | ||
BC049220.1 | - | 7 | 1542 | |||
BC053350.1 | - | 6 | 1640 | 5218414 | ||
BX648303.1 | - | 4 | 3339 | |||
CN286501.1 | - | 1 | 137 |