Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317955.1 | NP_001304884.1 | 4 | 682 | ||
NM_001317956.1 | NP_001304885.1 | 3 | 816 | |||
NM_001317957.1 | NP_001304886.1 | 2 | 643 | |||
NM_004508.3 | NP_004499.2 | 3 | 699 | |||
NR_134300.1 | 2 | 431 | ||||
NR_134301.1 | 2 | 526 | ||||
XM_011519457.1 | XP_011517759.1 | 3 | 1546 | |||
XM_017016191.1 | XP_016871680.1 | 2 | 1373 | |||
GenBank mRNA | AF271720.1 | - | 2 | 184 | ||
AK222875.1 | - | 3 | 219 | |||
AK303669.1 | - | 3 | 453 | |||
AK311110.1 | - | 2 | 75 | |||
BC005247.2 | - | 2 | 197 | |||
BC006999.2 | - | 2 | 197 | |||
BC019227.2 | - | 2 | 197 | |||
BC022418.2 | - | 2 | 197 | |||
BC025375.2 | - | 2 | 197 | |||
BC057827.1 | - | 4 | 670 | 30337530 | ||
BC107893.1 | - | 3 | 223 | |||
BE891119.1 | - | 3 | 503 | |||
BT006761.1 | - | 2 | 168 | |||
BX537663.1 | - | 2 | 3243 | |||
BX648472.1 | - | 2 | 1162 | |||
CD251816.1 | - | 3 | 328 | |||
CN481934.1 | - | 2 | 549 | |||
DA289206.1 | - | 2 | 517 | |||
DA792902.1 | - | 2 | 471 | |||
DA842794.1 | - | 3 | 151 | |||
X17025.1 | - | 3 | 218 |