Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005729.3 | NP_005720.1 | 6 | 1250 | ||
XM_005269379.2 | XP_005269436.1 | 6 | 1310 | |||
GenBank mRNA | AK296669.1 | - | 5 | 1167 | ||
AL832720.1 | - | 1 | 4248 | |||
AY358854.1 | - | 1 | 1385 | |||
BC005020.1 | - | 6 | 1236 | 3659599 | ||
BC013204.1 | - | 6 | 1227 | |||
BC110299.1 | - | 5 | 1151 | 5215228 |