Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001085354.2 | NP_001078823.1 | 7 | 1121 | ||
NM_024657.4 | NP_078933.3 | 7 | 1121 | |||
XM_005262190.3 | XP_005262247.1 | 6 | 1011 | |||
XM_006724691.2 | XP_006724754.1 | 5 | 755 | |||
XM_011531027.2 | XP_011529329.1 | 4 | 409 | |||
XM_017029844.1 | XP_016885333.1 | 6 | 1011 | |||
GenBank mRNA | AK056235.1 | - | 3 | 333 | ||
AK300305.1 | - | 6 | 696 | |||
AY927537.1 | - | LIT3543 AY927537 |
5 | 656 | ||
EF125209.1 | - | 7 | 846 | |||
EF125210.1 | - | 6 | 905 |