Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153712.4 | NP_714923.1 | 3 | 521 | ||
XM_005263599.3 | XP_005263656.1 | 3 | 560 | |||
XM_011510665.2 | XP_011508967.1 | 3 | 560 | |||
GenBank mRNA | AB071393.2 | - | 3 | 366 | ||
AK126549.1 | - | 1 | 13 | |||
BC036819.1 | - | 3 | 533 | 5767758 |