Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001218.4 | NP_001209.1 | 11 | 3463 | ||
NM_001293642.1 | NP_001280571.1 | 9 | 3250 | |||
NM_206925.2 | NP_996808.1 | 10 | 3430 | |||
NR_135511.1 | 10 | 3533 |