Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003892.1 | NP_001003892.1 | 1 | 40 | ||
XM_011539747.2 | XP_011538049.1 | 3 | 460 | |||
XM_017016176.1 | XP_016871665.1 | 2 | 333 | |||
GenBank mRNA | AY686755.1 | - | 1 | 40 | ||
BC137321.1 | - | 1 | 109 | 9021318 | ||
BC137322.1 | - | 1 | 109 | 9021319 |