Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEXA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000520.5 | NP_000511.2 | 14 | 2407 | ||
NM_001318825.1 | NP_001305754.1 | 14 | 2440 | |||
GenBank mRNA | M13520.1 | - | 14 | 1790 |