Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207063.1 | NP_001193992.1 | 5 | 848 | ||
NM_001207064.1 | NP_001193993.1 | 4 | 692 | |||
NM_001207065.1 | NP_001193994.1 | 3 | 487 | |||
NM_001207066.1 | NP_001193995.1 | 7 | 1110 | |||
NM_001338.4 | NP_001329.1 | 7 | 1110 | |||
XM_011529476.2 | XP_011527778.1 | 7 | 1000 | |||
XM_011529477.2 | XP_011527779.1 | 5 | 738 | |||
XM_011529478.2 | XP_011527780.1 | 5 | 738 | |||
XM_011529479.1 | XP_011527781.1 | 5 | 738 | |||
GenBank mRNA | AF124598.1 | - | 7 | 886 | ||
AK313526.1 | - | 7 | 973 | |||
AY072911.1 | - | 4 | 468 | |||
AY072912.1 | - | 5 | 624 | |||
BC003684.1 | - | 7 | 987 | 3463613 | ||
BC010536.1 | - | 7 | 961 | 3456544 | ||
BT019876.1 | - | 7 | 886 | |||
GU474812.1 | - | 6 | 763 | |||
U90716.1 | - | 7 | 886 | |||
Y07593.1 | - | 7 | 945 |