Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040442.1 | NP_001035532.1 | 4 | 430 | ||
NM_001130958.1 | NP_001124430.1 | 5 | 510 | |||
NM_001445.2 | NP_001436.1 | 2 | 345 | |||
GenBank mRNA | AI208231.1 | - | 4 | 430 | ||
BC022489.1 | - | 3 | 315 | 4792594 | ||
DQ132786.1 | - | 5 | 511 | |||
U19869.1 | - | 2 | 266 | |||
X90908.1 | - | 2 | 345 |