Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002166.4 | NP_002157.2 | 3 | 688 | ||
GenBank mRNA | BC030639.2 | - | 3 | 615 | 4820416 | |
D13891.1 | - | 3 | 601 | |||
M97796.1 | - | 3 | 613 |