Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003341.4 | NP_003332.1 | 2 | 294 | ||
NM_182666.2 | NP_872607.1 | 2 | 294 | |||
GenBank mRNA | AK314854.1 | - | 2 | 203 | ||
BC009139.1 | - | 2 | 218 | 3853408 | ||
BF698653.1 | - | 2 | 276 | |||
BI223271.1 | - | 2 | 189 | |||
DB478686.1 | - | 2 | 298 | |||
X92963.1 | - | 1 | 65 |