Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320715.1 | NP_001307644.1 | 3 | 1539 | ||
NM_003921.4 | NP_003912.1 | 3 | 1572 | |||
XM_011542397.2 | XP_011540699.1 | 3 | 1441 | |||
XM_011542398.2 | XP_011540700.1 | 3 | 1413 | |||
XM_011542399.2 | XP_011540701.1 | 3 | 1594 | |||
GenBank mRNA | AF057700.1 | - | 3 | 1267 | ||
AF082283.1 | - | 4 | 1726 | |||
AF100338.1 | - | 3 | 1267 | |||
AF134395.1 | - | 3 | 1265 | |||
AJ006288.1 | - | 4 | 1710 | |||
AK291346.1 | - | 3 | 1231 | |||
BC053617.1 | - | 3 | 1142 | 6143423 | ||
BX647306.1 | - | 4 | 1120 |