Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004705.3 | NP_004696.2 | 5 | 2138 | ||
NR_130898.1 | 6 | 2233 | ||||
GenBank mRNA | AF081567.1 | - | 5 | 2009 | ||
AK001522.1 | - | 1 | 577 | |||
AK291843.1 | - | 5 | 2010 | |||
AK299594.1 | - | 3 | 1769 | |||
AL049970.1 | - | 1 | 1542 | |||
BC021992.1 | - | 1 | 428 | |||
BC110304.1 | - | 5 | 2115 | 5222582 | ||
BC117138.1 | - | 5 | 2067 | 40125689 | ||
BC117140.1 | - | 5 | 2067 | 40125691 | ||
BX641144.1 | - | 2 | 5609 |