Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53I3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206802.2 | NP_001193731.1 | 4 | 920 | ||
NM_004881.4 | NP_004872.2 | 5 | 2001 | |||
NM_147184.3 | NP_671713.1 | 6 | 1602 | |||
XM_005264650.2 | XP_005264707.1 | 6 | 1474 | |||
XM_006712150.2 | XP_006712213.1 | 5 | 1405 | |||
GenBank mRNA | AF010309.1 | - | 5 | 1639 | ||
AK223382.1 | - | 5 | 1838 | |||
AK297579.1 | - | 5 | 1442 | |||
BC000474.2 | - | 6 | 1172 | 2961571 | ||
BC018819.2 | - | 6 | 1172 | |||
CN285566.1 | - | 2 | 443 |