Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278309.1 | NP_001265238.1 | 5 | 1405 | ||
NM_006422.3 | NP_006413.3 | 5 | 1383 | |||
XM_005253664.2 | XP_005253721.1 | 5 | 1315 | |||
XM_011520908.1 | XP_011519210.1 | 4 | 1519 | |||
XM_011520909.1 | XP_011519211.1 | 3 | 996 | |||
XM_011520910.2 | XP_011519212.1 | 3 | 1451 | |||
GenBank mRNA | AF087003.1 | - | 4 | 1020 | ||
AF093408.1 | - | 4 | 1087 | |||
AK074722.1 | - | 1 | 476 | |||
AK292451.1 | - | 5 | 1383 | |||
BC047535.1 | - | 4 | 1067 | 5267552 | ||
EU794681.1 | - | 4 | 1087 | |||
U85715.1 | - | 4 | 1060 |