Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006699.3 | NP_006690.1 | 13 | 2624 | ||
GenBank mRNA | AF027156.1 | - | 13 | 2423 | ||
BC063300.1 | - | 13 | 2622 | 5263292 |