Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171191.1 | NP_001164662.1 | 6 | 800 | ||
NM_001171192.1 | NP_001164663.1 | 8 | 1018 | |||
NM_001171193.1 | NP_001164664.1 | 7 | 904 | |||
NM_017711.3 | NP_060181.2 | 8 | 1018 | |||
XM_011530977.1 | XP_011529279.1 | 5 | 534 | |||
XM_017029614.1 | XP_016885103.1 | 7 | 999 | |||
GenBank mRNA | AB048363.1 | - | 8 | 908 | ||
AK000214.1 | - | 8 | 730 | |||
AK097293.1 | - | 4 | 296 | |||
AK299255.1 | - | 6 | 628 | |||
AK301025.1 | - | 8 | 842 | |||
AK307778.1 | - | 8 | 719 | |||
AK310090.1 | - | 4 | 658 | |||
AK316035.1 | - | 7 | 636 | |||
AY358986.1 | - | 8 | 713 | |||
BC032009.1 | - | 8 | 715 | 4747934 |