Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDLIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114107.4 | NP_001107579.1 | 7 | 1200 | ||
NM_001257962.1 | NP_001244891.1 | 7 | 1080 | |||
NM_001257963.1 | NP_001244892.1 | 5 | 843 | |||
NM_014476.5 | NP_055291.2 | 8 | 1344 | |||
NR_047562.1 | 6 | 1012 | ||||
GenBank mRNA | AF002280.1 | - | 8 | 1264 | ||
AF002282.1 | - | 7 | 1120 | |||
AF039018.1 | - | 8 | 1290 | |||
AK129863.1 | - | 6 | 881 | |||
AK222966.1 | - | 8 | 1248 | |||
BC027870.1 | - | 7 | 1127 | 5205215 | ||
BX647263.1 | - | 4 | 4721 | |||
CD625728.1 | - | 3 | 547 | |||
CR739525.1 | - | 1 | 301 |