Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130767.2 | NP_570123.1 | 14, 15 | 1568 | ||
XM_006714531.2 | XP_006714594.1 | 13, 14 | 1449 | |||
XM_017009045.1 | XP_016864534.1 | 14, 15 | 1533 | |||
XM_017009046.1 | XP_016864535.1 | 13, 14 | 1449 | |||
XM_017009047.1 | XP_016864536.1 | 13, 14 | 1471 | |||
XM_017009048.1 | XP_016864537.1 | 10, 11 | 1245 | |||
GenBank mRNA | AB078619.1 | - | 14, 15 | 1536 | ||
AK122960.1 | - | 14, 15 | 1533 | |||
BC075010.2 | - | 14, 15 | 1534 | 30915616 | ||
BC075011.2 | - | 14, 15 | 1529 | 30915403 |