Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP8B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178002.2 | NP_001171473.1 | 29 | 4385 | ||
NM_138813.3 | NP_620168.1 | 29 | 4476 | |||
NR_047593.2 | 29 | 4620 | ||||
XM_006722654.2 | XP_006722717.1 | 29 | 4506 | |||
XM_006722655.2 | XP_006722718.1 | 29 | 4585 | |||
XM_006722656.2 | XP_006722719.1 | 29 | 4394 | |||
XM_011527707.1 | XP_011526009.1 | 29 | 4479 | |||
XM_011527708.1 | XP_011526010.1 | 29 | 4386 | |||
XM_011527709.1 | XP_011526011.1 | 28 | 4293 | |||
XM_011527710.2 | XP_011526012.1 | 23 | 3715 | |||
XM_011527712.1 | XP_011526014.1 | 14 | 2508 | |||
GenBank mRNA | AK057452.1 | - | 6 | 1336 | ||
AK058189.1 | - | 2 | 701 | |||
AK125969.1 | - | 29 | 4620 | |||
BC035162.2 | - | 29 | 4471 | 5265541 |