Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039667.2 | NP_001034756.1 | 6 | 1208 | ||
NM_139314.2 | NP_647475.1 | 7 | 1322 | |||
XM_005272484.3 | XP_005272541.1 | 7 | 1374 | |||
XM_005272485.3 | XP_005272542.1 | 6 | 1260 | |||
GenBank mRNA | AB056477.1 | - | 7 | 1296 | ||
AF153606.1 | - | 7 | 1277 | |||
AF169312.1 | - | 7 | 1191 | |||
AF202636.1 | - | 7 | 1299 | |||
AK222489.1 | - | 7 | 1294 | |||
AK222524.1 | - | 7 | 1292 | |||
AK303269.1 | - | 8 | 1223 | |||
AW954426.1 | - | 1 | 13 | |||
AY358275.1 | - | 7 | 1290 | |||
BC023647.2 | - | 7 | 1266 | 5088323 |