Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145059.2 | NP_659496.2 | 24 | 3547 | ||
XM_006721162.1 | XP_006721225.1 | 23 | 3477 | |||
XM_011522928.1 | XP_011521230.1 | 24 | 3725 | |||
XM_011522929.1 | XP_011521231.1 | 24 | 3590 | |||
XM_017023012.1 | XP_016878501.1 | 24 | 3574 | |||
XM_017023013.1 | XP_016878502.1 | 21 | 3370 | |||
XM_017023014.1 | XP_016878503.1 | 19 | 3502 | |||
GenBank mRNA | AJ441184.1 | - | 24 | 3546 | ||
AK056456.1 | - | 24 | 3546 | |||
AK074168.1 | - | 22 | 4078 | |||
AK092025.1 | - | 16 | 2696 | |||
AK096727.1 | - | 16 | 2753 | |||
AK123470.1 | - | 3 | 2291 | |||
AK128387.1 | - | 24 | 3565 | |||
AY358187.1 | - | 2 | 1125 | |||
BC013735.1 | - | 22 | 3333 | |||
BC032542.1 | - | 24 | 3553 | 5534376 |