Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153213.3 | NP_694945.2 | 16 | 2800 | ||
XM_011540706.2 | XP_011539008.1 | 17 | 2964 | |||
GenBank mRNA | AK091281.1 | - | 9 | 1936 | ||
AK297707.1 | - | 12 | 2299 | |||
AL137736.1 | - | 11 | 1556 | |||
BC012982.2 | - | 15 | 2376 | |||
BC040640.1 | - | 16 | 2800 | 3847806 |