Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003102.2 | NP_003093.2 | 2 | 317 | ||
GenBank mRNA | AK313935.1 | - | 2 | 298 | ||
BC014418.1 | - | 2 | 182 | 4644224 | ||
CR541853.1 | - | 1 | 112 | |||
DA668533.1 | - | 2 | 302 | |||
J02947.1 | - | 2 | 181 |