Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC39A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280557.1 | NP_001267486.1 | 2 | 99 | ||
NM_017767.2 | NP_060237.2 | 9 | 1744 | |||
NM_130849.3 | NP_570901.2 | 10 | 1600 | |||
GenBank mRNA | AK000334.1 | - | 9 | 1724 | ||
AK000489.1 | - | 2 | 94 | |||
AK000695.1 | - | 9 | 1659 | |||
AK025537.1 | - | 10 | 1594 | |||
AK056900.1 | - | 2 | 93 | |||
AK290563.1 | - | 10 | 1548 | |||
BC001688.2 | - | 2 | 71 | |||
BC062625.1 | - | 10 | 1580 | 5203858 | ||
BE798833.1 | - | 1 | 230 | |||
KU178707.1 | - | 10 | 1494 | |||
KU178708.1 | - | 9 | 1212 |