Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001442.2 | NP_001433.1 | 2 | 223 | ||
GenBank mRNA | BC003672.1 | - | 2 | 218 | 3683235 | |
BQ880795.1 | - | 2 | 155 | |||
BT006809.1 | - | 2 | 152 | |||
CD000452.1 | - | 2 | 224 | |||
CR456903.1 | - | 2 | 152 | |||
FJ224319.1 | - | 2 | 214 | |||
J02874.1 | - | 2 | 214 |