Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287060.1 | NP_001273989.1 | 4 | 1112 | ||
NM_001287061.1 | NP_001273990.1 | 2 | 285 | |||
NM_001287062.1 | NP_001273991.1 | 3 | 421 | |||
NM_014504.2 | NP_055319.1 | 2 | 226 | |||
GenBank mRNA | AJ250042.1 | - | 2 | 149 | ||
AK056864.1 | - | 4 | 1039 | |||
AK223329.1 | - | 2 | 149 | |||
AK290678.1 | - | 3 | 344 | |||
AK303006.1 | - | 2 | 282 | |||
BC015330.1 | - | 2 | 172 | 4420279 | ||
BT007107.1 | - | 1 | 72 | |||
CN305879.1 | - | 4 | 335 | |||
DA196847.1 | - | 2 | 226 | |||
DC402344.1 | - | 2 | 287 | |||
DQ230533.1 | - | 1 | 72 | |||
DQ230534.1 | - | 1 | 72 | |||
DQ230535.1 | - | 1 | 72 |