Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEX3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032246.4 | NP_115622.2 | 2 | 1423 | ||
GenBank mRNA | AB095929.1 | - | 1 | 2235 | ||
AK094145.1 | - | 1 | 656 | |||
AK094633.1 | - | 1 | 87 | |||
AK131424.1 | - | 2 | 1300 | |||
AL136778.1 | - | 1 | 77 | |||
AY950678.1 | - | 2 | 985 | |||
BC036211.1 | - | 2 | 1421 | 5266259 | ||
BC111545.1 | - | 2 | 1399 | 5742971 |