Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024736.1 | NP_001019907.1 | 4 | 836 | ||
NM_025240.2 | NP_079516.1 | 4 | 836 | |||
XM_005254700.4 | XP_005254757.1 | 4 | 954 | |||
XM_011522095.2 | XP_011520397.1 | 4 | 825 | |||
XM_011522096.2 | XP_011520398.1 | 4 | 765 | |||
XM_017022638.1 | XP_016878127.1 | 4 | 754 | |||
GenBank mRNA | AF302102.1 | - | 3 | 602 | ||
AJ583695.1 | - | 3 | 645 | |||
AK074849.1 | - | 4 | 726 | |||
AK074997.1 | - | 2 | 332 | |||
AK075549.1 | - | 4 | 764 | |||
AK291721.1 | - | 4 | 673 | |||
AK292711.1 | - | 3 | 597 | |||
AK296358.1 | - | 4 | 554 | |||
AL360136.1 | - | 1 | 164 | |||
AY358343.1 | - | 4 | 753 | |||
BC062581.1 | - | 4 | 836 | 5201491 | ||
BQ278722.1 | - | 419 |