Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015.4 | NP_001006.1 | 3 | 288 | ||
GenBank mRNA | AK311809.1 | - | 3 | 223 | ||
AV759412.2 | - | 3 | 229 | |||
BC007283.1 | - | 3 | 247 | 3343839 | ||
BC007603.1 | - | 3 | 247 | 3350204 | ||
BC007945.2 | - | 3 | 248 | 4297932 | ||
BC010028.2 | - | 3 | 247 | 3357773 | ||
BC016378.1 | - | 1 | 202 | 4667201 | ||
BC018829.2 | - | 3 | 247 | |||
BC070224.1 | - | 1 | 199 | 4809182 | ||
BC100025.1 | - | 3 | 298 | 30565084 | ||
BF239060.1 | - | 3 | 275 | |||
BI550467.1 | - | 3 | 224 | |||
DA879881.1 | - | 3 | 288 | |||
X06617.1 | - | 3 | 211 |