Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005.4 | NP_000996.2 | 1, 2 | 91 | ||
NM_001256802.1 | NP_001243731.1 | 1, 2 | 91 | |||
NM_001260506.1 | NP_001247435.1 | 1, 2 | 91 | |||
GenBank mRNA | AB062288.1 | - | 1, 2 | 27 | ||
AK129794.1 | - | 1, 2 | 63 | |||
AK130960.1 | - | 1, 2 | 64 | |||
AK223048.1 | - | 1, 2 | 40 | |||
AK313051.1 | - | 1, 2 | 60 | |||
BC003137.1 | - | 1, 2 | 40 | 2906158 | ||
BC003577.1 | - | 1, 2 | 32 | |||
BC013196.1 | - | 1, 2 | 32 | |||
BC013231.1 | - | 1, 2 | 52 | |||
BC034149.1 | - | 1, 2 | 64 | 4665438 | ||
BC071917.1 | - | 1, 2 | 43 | 6297012 | ||
BC100284.1 | - | 1, 2 | 52 | 6160514 | ||
BM831460.1 | - | 1, 2 | 91 | |||
BM928330.1 | - | 1, 2 | 73 | |||
BQ645330.1 | - | 51 | ||||
S42658.1 | - | 1, 2 | 42 | |||
U14990.1 | - | 1, 2 | 34 | |||
U14991.1 | - | 1, 2 | 34 | |||
U14992.1 | - | 1, 2 | 34 | |||
X55715.1 | - | 1, 2 | 56 |