Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR1F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012360.1 | NP_036492.1 | 1 | 886 | ||
XM_011522506.2 | XP_011520808.1 | 4 | 1333 | |||
XM_011522507.2 | XP_011520809.1 | 2 | 1756 | |||
GenBank mRNA | BC069335.1 | - | 1 | 886 | 7262265 | |
Y14442.1 | - | 1 | 886 |