Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030001.2 | NP_001025172.1 | 2 | 221 | ||
NM_001032.4 | NP_001023.1 | 2 | 221 | |||
GenBank mRNA | AK026649.1 | - | 1 | 181 | ||
AK307833.1 | - | 2 | 182 | |||
BC015974.1 | - | 2 | 185 | 4105424 | ||
BC032813.1 | - | 1 | 156 | 5217083 | ||
BC035313.1 | - | 156 | 3868721 | |||
BF128896.1 | - | 2 | 128 | |||
BQ433530.1 | - | 2 | 165 | |||
BU942244.1 | - | 2 | 224 | |||
L31610.1 | - | 2 | 162 | |||
U14973.1 | - | 2 | 181 |