Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BANF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143985.1 | NP_001137457.1 | 3 | 1004 | ||
NM_003860.3 | NP_003851.1 | 3 | 1061 | |||
XM_017018514.1 | XP_016874003.1 | 3 | 1054 | |||
XM_017018515.1 | XP_016874004.1 | 3 | 594 | |||
GenBank mRNA | AF044773.1 | - | 3 | 654 | ||
AF068235.1 | - | 3 | 1060 | |||
BC005942.1 | - | 3 | 640 | 4074168 | ||
BC107702.1 | - | 3 | 737 | 6495215 |