Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130678.1 | NP_001124150.1 | 7 | 924 | ||
NM_001130679.1 | NP_001124151.1 | 8 | 2360 | |||
NM_001968.3 | NP_001959.1 | 7 | 2267 | |||
XM_006714126.3 | XP_006714189.1 | 8 | 1970 | |||
GenBank mRNA | AK300982.1 | - | 7 | 924 | ||
BC012611.1 | - | 7 | 760 | 4282826 | ||
BC035166.1 | - | 7 | 2275 | 5265600 | ||
BC043226.1 | - | 7 | 1606 | 5295521 | ||
BC107715.1 | - | 7 | 761 | 6716042 | ||
M15353.1 | - | 7 | 762 |