Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006097.4 | NP_006088.2 | 4 | 551 | ||
NM_181526.2 | NP_852667.1 | 3 | 389 | |||
GenBank mRNA | AF176042.1 | - | 3 | 487 | ||
AF176043.1 | - | 2 | 325 | |||
AK130879.1 | - | 4 | 516 | |||
BC002648.2 | - | 3 | 328 | 3608316 | ||
BQ953325.1 | - | 541 | ||||
CR456843.1 | - | 3 | 447 | |||
D82057.1 | - | 4 | 519 |