Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033034.2 | NP_149023.2 | 2 | 600 | ||
NM_033092.2 | NP_149083.2 | 2 | 600 | |||
NM_033093.2 | NP_149084.2 | 2 | 452 | |||
XM_005253183.2 | XP_005253240.1 | 2 | 452 | |||
XM_005253184.2 | XP_005253241.1 | 2 | 586 | |||
XM_006718358.2 | XP_006718421.1 | 2 | 586 | |||
XM_011520426.2 | XP_011518728.1 | 2 | 586 | |||
XM_011520427.1 | XP_011518729.1 | 2 | 586 | |||
XM_017018460.1 | XP_016873949.1 | 2 | 586 | |||
XM_017018461.1 | XP_016873950.1 | 2 | 452 | |||
XM_017018462.1 | XP_016873951.1 | 2 | 586 | |||
XM_017018463.1 | XP_016873952.1 | 2 | 452 | |||
GenBank mRNA | AB362928.1 | - | 1 | 297 | ||
AF220025.1 | - | 1 | 359 | |||
AF220026.1 | - | 359 | ||||
AF220027.1 | - | 1 | 359 | |||
AF220028.1 | - | 1 | 359 | |||
AF220029.1 | - | 1 | 359 | |||
AK027593.1 | - | 2 | 554 | |||
AY625000.1 | - | 1 | 297 | |||
BC020770.1 | - | 3 | 723 | |||
BC021258.2 | - | 2 | 533 | 4122548 | ||
BX647152.1 | - | 3 | 489 | |||
CN334837.1 | - | 1 | 260 | |||
DA475412.1 | - | 2 | 452 | |||
DQ288685.1 | - | 1 | 297 | |||
JF928461.1 | - | 1 | 311 | |||
JF928462.1 | - | 1 | 311 |