Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242374.1 | NP_001229303.1 | 3 | 1334 | ||
NM_001242375.1 | NP_001229304.1 | 3 | 919 | |||
NM_015391.3 | NP_056206.1 | 3 | 700 | |||
GenBank mRNA | AK001285.1 | - | 3 | 1331 | ||
AK026550.1 | - | 3 | 654 | |||
AL050051.1 | - | 3 | 919 | |||
BC005398.1 | - | 3 | 666 | 3828623 | ||
BU852706.1 | - | 3 | 680 | |||
BX537696.1 | - | 4 | 1120 |